Learn More
Cayman Chemical BIsIndolylmaleImIde II 1mg
SDP
Supplier:  Cayman Chemical 110201 MG
Encompass_Preferred
A general inhibitor of all PKC subtypes with structural similarity to the nonspecific PKC inhibitor staurosporine; inhibits PDK1 (IC50 = 14 μM) and PKA (IC50 = 2.94 μM)
Catalog No. 50-295-1625