Learn More
TARGETMOL CHEMICALS INC Bisindolylmaleimide III 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T7762810MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Bisindolylmaleimide III is a potent and selective inhibitor of protein kinase C (PKC), specifically interacting with PKCalpha or ribosomal S6 protein kinase 1 upon activation of these kinases. Purity 98.6%
Catalog No. 50-287-6617