Learn More
Medchemexpress LLC Bisindolylmaleimide III | 137592-43-9 | 99.0% | 384.43 g/mol | C23H20N4O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11739310MG
Encompass_Preferred
Bisindolylmaleimide III is a small-molecule research reagent and a potent, selective inhibitor of protein kinase C (PKC), with reported activity against PKCα and interactions with ribosomal S6 protein kinase 1. It is supplied for laboratory research use and is characterized by a molecular formula of C23H20N4O2, molecular weight of 384.43 g/mol, and a manufacturer-stated purity of approximately 98.95%.
  • Potent and selective PKC inhibitor.
  • Characterized by molecular weight 384.43 g/mol and formula C23H20N4O2.
  • High purity suitable for biochemical and cell signaling studies.
  • Supplied in small-scale quantities for research use.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-27520