Learn More
Cayman Chemical BIsIndolylmaleImIde III 5mg
SDP
Supplier:  Cayman Chemical 110725 MG
Encompass_Preferred
A protein kinase C inhibitor with structural similarity to staurosporine; inhibits PKCα (93% kinase activity inhibited at 1 μM ), PDK1 (IC50 =3.8 μM), S6K1, MAPKAP-K1, RSK2 and MSK1
Catalog No. 50-295-1597