Learn More
TARGETMOL CHEMICALS INC BISINDOLYLMALEIMIDE IV 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T765910MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bisindolylmaleimide IV is a protein kinase C (PKC) cell permeable inhibitor( IC50 0.10 - 0.55 uM)purity: 98%
Catalog No. 50-257-9164