Learn More
Medchemexpress LLC Bisindolylmaleimide IV | 119139-23-0 | MFCD00236432 | 99.7% | 327.34 g/mol | C20H13N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10825410MG
Encompass_Preferred
Bisindolylmaleimide IV is a small-molecule inhibitor of protein kinase C (PKC) and a selective inhibitor of human cytomegalovirus (HCMV) replication in cell culture. Supplied as a research reagent, it has characterized solubility and storage profiles and is intended for in vitro pharmacology and analytical applications.
  • Potent inhibitor of protein kinase C (PKC).
  • Selective inhibitor of human cytomegalovirus replication in cell culture.
  • High purity (99.71%) suitable for research use.
  • Soluble in DMSO (125 mg/mL); clear solution.
  • Available in small research quantities and 10 mM DMSO solutions.
  • Defined storage conditions for powder and solvent preparations.
Catalog No. 50-002-42541