Learn More
Medchemexpress LLC Bisindolylmaleimide IV | 119139-23-0 | MFCD00236432 | 99.7% | 327.34 g·mol⁻¹ | C20H13N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10825450MG
Encompass_Preferred
Bisindolylmaleimide IV (Arcyriarubin A) is a research-grade small-molecule inhibitor of protein kinase C used in biochemical and cellular studies. It exhibits submicromolar activity against PKC, shows activity against PKA, and has demonstrated suppression of human cytomegalovirus replication in cell culture. The compound is supplied as a solid powder or as a 10 mM solution in DMSO for laboratory applications.
  • Potent PKC inhibition (IC50 0.1-0.55 μM).
  • Also inhibits PKA (IC50 3.1-11.8 μM) and HCMV replication (IC50 0.2 μM).
  • High reported purity suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO for convenience.
  • Supplied in multiple small-scale pack sizes for laboratory use.
Catalog No. 50-002-43961