Learn More
AdipoGen Bisindolylmaleimide IV
SDP
Supplier:  AdipoGen AGCR10152M005
Encompass_Preferred
Chemical. CAS 119139-23-0. Formula C20H13N3O2. MW 327.3. Cell permeable protein kinase C PKC inhibitor. Inhibits also protein kinase A PKA.
Catalog No. 50-256-2359