Learn More
Medchemexpress LLC Bisindolylmaleimide IV (Arcyriarubin A) | 119139-23-0 | MFCD00236432 | 99.7% | 327.34 g·mol-1 | C20H13N3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10825425MG
Encompass_Preferred
Bisindolylmaleimide IV (Arcyriarubin A) is a small-molecule research reagent that potently inhibits protein kinase C (PKC) and reduces human cytomegalovirus replication in cell culture. Supplied as a high-purity solid for in vitro assays, prepare solutions in appropriate solvents and follow safety guidance for handling and storage.
  • Potent protein kinase C inhibitor with PKC IC50 ≈ 0.1-0.55 μM.
  • Inhibits human cytomegalovirus replication (IC50 ≈ 0.2 μM).
  • High purity (99.7%) suitable for in vitro research.
  • Small-molecule solid; molecular weight 327.34 g·mol-1, formula C20H13N3O2.
  • Soluble in DMSO and common organic solvents for assay preparation.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-41963