Learn More
Selleck Chemical LLC Bisindolylmaleimide IX
SDP
Supplier:  Selleck Chemical LLC S720750MG
Encompass_Preferred
Bisindolylmaleimide IX (Ro 31-8220 Mesylate) is a pan-PKC inhibitor with IC50 of 5 nM 24 nM 14 nM 27 nM and 24 nM for PKC- PKC- I PKC- II PKC- and PKC- respectively and also shows potent inhibition against MAPKAP-K1b MSK1 GSK3 and S6K1
Catalog No. 50-000-04335