Learn More
AdipoGen Bisindolylmaleimide V
SDP
Supplier:  AdipoGen AGCR10023M005
Encompass_Preferred
Chemical. CAS 113963-68-1. Formula C21H15N3O2. MW 341.4. Negative control for protein kinase C PKC inhibitors. S6K inhibitor. Necrosis inhibitor.
Catalog No. 50-256-2392