Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | 97.8% | 341.36 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11240010MG
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies. It blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo.
  • Cell-permeable negative control.
  • IC50 value over 100 μM for protein kinase C inhibition.
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo.
  • IC50 of 8 μM for S6K inhibition.
Catalog No. 50-004-38481