Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | 97.8% | 341.36 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY112400
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies with an IC50 value over 100 μM. It blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) *in vivo* with an IC50 of 8 μM. This compound is suitable for laboratory research involving protein kinase inhibition and S6K signaling pathways.
  • Cell-permeable negative control for protein kinase C inhibition studies
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K)
  • Useful for S6K signaling research
Catalog No. 50-004-39693