Learn More
Medchemexpress LLC Bisindolylmaleimide V | 113963-68-1 | 97.8% | 341.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11240050MG
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, with an IC50 value exceeding 100 μM. It blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo, with an IC50 of 8 μM. This product is for research use only.
  • Cell-permeable negative control for protein kinase C inhibition studies
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo
  • Appears as a solid with an orange to red color
Catalog No. 50-004-36433