Learn More
Cayman Chemical BIsIndolylmaleImIde V 1mg
SDP
Supplier:  Cayman Chemical 133001 MG
Encompass_Preferred
A weak inhibitor of PKC demonstrating an IC50 value >100 µM; blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo with an IC50 value of 8 µM
Catalog No. 50-295-2899