Learn More
Medchemexpress LLC Bisindolylmaleimide V (BIM V) | 113963-68-1 | 97.8% | 341.36 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY112400
Encompass_Preferred
Bisindolylmaleimide V is a cell-permeable negative control for protein kinase C inhibition studies, with an IC50 value over 100 μM. It blocks the activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo with an IC50 of 8 μM. It is for research use only.
  • Cell-permeable negative control for protein kinase C inhibition studies
  • Blocks activation of mitogen-stimulated protein kinase p70s6k/p85s6k (S6K) in vivo
  • Intended for research use only
Catalog No. 50-004-39787