Learn More
TARGETMOL CHEMICALS INC Bisindolylmaleimide VIII aceta
SDP
Supplier:  TARGETMOL CHEMICALS INC T1054910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Bisindolylmaleimide VIII acetate (Ro 31-7549 acetate) is a potent and selective PKC inhibitor (IC50 158 nM for rat brain PKC) with IC50 values of 53, 195, 163, 213, and 175 nM for PKC-alpha, PKC-betaI, PKC-betaII, PKC-gamma, and PKC-, respectively. Purity 99%
Catalog No. 50-288-0731