Learn More
Medchemexpress LLC Bisindolylmaleimide VIII acetate | 138516-31-1 | 99.68% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY129624A50MG
Encompass_Preferred
Bisindolylmaleimide VIII acetate (Ro 31-7549 acetate) is a potent and selective protein kinase C (PKC) inhibitor with an IC50 of 158 nM for rat brain PKC. It has IC50s of 53, 195, 163, 213, and 175 nM for PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε, respectively. Bisindolylmaleimide VIII acetate facilitates Fas-mediated apoptosis and inhibits T cell-mediated autoimmune diseases. It is for research use only and not sold to patients.
  • Potent and selective protein kinase C (PKC) inhibitor
  • Facilitates Fas-mediated apoptosis
  • Inhibits T cell-mediated autoimmune diseases
  • For research use only
Catalog No. 50-003-42367