Learn More
Medchemexpress LLC Bisindolylmaleimide VIII acetate | 138516-31-1 | 99.7% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY129624A
Encompass_Preferred
Bisindolylmaleimide VIII acetate (Ro 31-7549 acetate) is a potent and selective protein kinase C (PKC) inhibitor, with an IC50 of 158 nM for rat brain PKC. It specifically inhibits various PKC isoforms including PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε. This compound facilitates Fas-mediated apoptosis and inhibits T cell-mediated autoimmune diseases. It is supplied as a brown to reddish-brown solid with a molecular weight of 458.51 and chemical formula C26H26N4O4. This product is for research use only.
  • Potent and selective protein kinase C (PKC) inhibitor
  • Inhibits PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε isoforms
  • Facilitates Fas-mediated apoptosis
  • Inhibits T cell-mediated autoimmune diseases
  • High purity of 99.7%
  • Supplied as a solid with a brown to reddish-brown color
Catalog No. 50-003-42742