Learn More
Medchemexpress LLC Bisindolylmaleimide VIII acetate | 138516-31-1 | 99.7% | 458.51 g/mol | C26H26N4O4 | 5 MG

Supplier: Medchemexpress LLC HY129624A5MG
Bisindolylmaleimide VIII acetate is a bisindolylmaleimide derivative used as a research reagent to inhibit protein kinase C (PKC) activity in biochemical and cellular studies. It is used to probe PKC-mediated signaling and apoptosis pathways and is supplied in small milligram-scale vials for laboratory use.
- High purity of 99.7%.
- Molecular weight 458.51 g/mol.
- Molecular formula C26H26N4O4.
- Brown to reddish brown solid appearance.
- Potent inhibitor of protein kinase C and multiple PKC isoforms.
- Suitable for enzymatic and cell-based assays studying apoptosis.
- Available in small milligram vials (5 mg-50 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.