Learn More
Medchemexpress LLC Bisindolylmaleimide VIII acetate (Ro 31-7549 acetate) | 138516-31-1 | 99.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY129624A10MG
Encompass_Preferred
Bisindolylmaleimide VIII acetate (Ro 31-7549 acetate) is a potent and selective protein kinase C (PKC) inhibitor. It demonstrates inhibitory effects on various PKC isoforms including PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε. This compound is known to facilitate Fas-mediated apoptosis and has shown promise in inhibiting T cell-mediated autoimmune diseases.
  • Potent and selective protein kinase C (PKC) inhibitor
  • Inhibits PKC-α, PKC-βI, PKC-βII, PKC-γ, and PKC-ε isoforms
  • Facilitates Fas-mediated apoptosis
  • Inhibits T cell-mediated autoimmune diseases
Catalog No. 50-003-40173