Learn More
Medchemexpress LLC HY-108136A 10mg Medchemexpress, Bisindolylmaleimide X (hydrochloride) CAS:145317-11-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108136A10MG
Encompass_Preferred
Medchemexpress, HY-108136A 10mg Bisindolylmaleimide X (HCl) CAS:145317-11-9 Bisindolylmaleimide X hydrochloride (BIM-X hydrochloride) is a potent and selective protein kinase C (PKC) inhibitor. Bisindolylmaleimide X hydrochloride is a potent cyclin-dependent kinase 2 (CDK2) antagonist with an IC50 of 200 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8509