Learn More
Medchemexpress LLC Bisindolylmaleimide X hydrochloride | 145317-11-9 | MFCD04972010 | >90.0% | 460.96 | C26H25ClN4O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY108136A10MM 1ML
Encompass_Preferred
Bisindolylmaleimide X hydrochloride is a cell-permeable, reversible, ATP-competitive protein kinase C (PKC) inhibitor commonly used in biochemical and cell biology research to study kinase activity and signal transduction. Supplied as a ready-to-use solution, it facilitates rapid setup of in vitro assays; consult the supplier datasheet for handling and storage instructions.
  • Potent protein kinase C inhibitor for biochemical assays.
  • Cell-permeable and reversible mechanism of action.
  • Provided as a ready-to-use high-concentration solution for immediate use.
  • Suitable for kinase inhibition and signaling pathway studies.
  • Follow supplier safety and storage recommendations for stability.
Catalog No. 50-002-42462