Learn More
Medchemexpress LLC Bisindolylmaleimide X hydrochloride | 145317-11-9 | MFCD04972010 | 98.9% | 460.96 g/mol | C26H25ClN4O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108136A50MG
Encompass_Preferred
Bisindolylmaleimide X hydrochloride is a small-molecule kinase inhibitor supplied as a solid for research use. It acts primarily as a potent and selective protein kinase C (PKC) inhibitor and also antagonizes cyclin-dependent kinase 2 (CDK2). The material is provided at high purity for biochemical and cell-based assays.
  • Potent protein kinase C (PKC) inhibitor
  • Antagonizes cyclin-dependent kinase 2 with reported IC50 ≈ 200 nM
  • High purity suitable for biochemical and cell-based studies
  • Solid form, available in small laboratory pack sizes
  • Useful for kinase signaling and pharmacology research
Catalog No. 50-002-43036