Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Bismuth Chloride, 10g

Supplier:  ALADDIN SCIENTIFIC CORPORATION B10034010G

Encompass_Preferred

Bismuth Chloride PrimorTrace 99 99% metals basisCas-7787-60-2

Catalog No. 50-000-82499


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.