Learn More
Strem, An Ascensus Company CAS# 7787-64-6. 5g. Bismuth(III) iodide (99.999%-Bi) PURATREM. MFCD00010894
SDP
Supplier:  Strem, An Ascensus Company 9383175G
Encompass
CAS# 7787-64-6. 5g. Bismuth(III) iodide (99.999%-Bi) PURATREM. MFCD00010894. Molecular Weight: 589.69. Molecular Formula: BiI3. Color/form: gray pwdr. Strem# 93-8317. http://www.strem.com/catalog/v/93-8317/
Catalog No. 50-901-16542