Learn More
Strem, An Ascensus Company CAS# 34364-26-6. 10g. Bismuth(III) neodecanoate, superconductor grade ~60% in neodecanoic acid (15-20% Bi). MFCD00070469
SDP
Supplier:  Strem, An Ascensus Company 83700010G
Encompass
CAS# 34364-26-6. 10g. Bismuth(III) neodecanoate, superconductor grade ~60% in neodecanoic acid (15-20% Bi). MFCD00070469. Molecular Weight: 722.71. Molecular Formula: Bi(OOCC9H19-neo)3. Color/form: viscous liq. Strem# 83-7000. http://www.strem.com/catalog/v/83-7000/
Catalog No. 50-901-13552