Learn More
Strem, An Ascensus Company CAS# 1304-76-3. 25g. Bismuth(III) oxide (99.9998%-Bi) PURATREM. MFCD00003462
SDP
Supplier:  Strem, An Ascensus Company 83402525G
Encompass
CAS# 1304-76-3. 25g. Bismuth(III) oxide (99.9998%-Bi) PURATREM. MFCD00003462. Molecular Weight: 465.96. Molecular Formula: Bi2O3. Color/form: yellow pwdr. Strem# 83-4025. http://www.strem.com/catalog/v/83-4025/
Catalog No. 50-901-13550