Learn More
Strem, An Ascensus Company CAS# 12048-51-0. 100g. Bismuth(III) titanate, 98%. MFCD00136014
SDP
Supplier:  Strem, An Ascensus Company 938312100G
Encompass
CAS# 12048-51-0. 100g. Bismuth(III) titanate, 98%. MFCD00136014. Molecular Weight: 625.76. Molecular Formula: Bi2Ti2O7. Color/form: white pwdr. Strem# 93-8312. http://www.strem.com/catalog/v/93-8312/
Catalog No. 50-901-16533