Learn More
Medchemexpress LLC Bisoctrizole | 103597-45-1 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB08975G
Encompass_Preferred
Bisoctrizole is a broad-spectrum UVA/UVB absorber from the benzotriazole class. It works by absorbing UV light through its hydroxyphenyl groups, converting light energy into heat to prevent UV-induced damage to materials or skin. It is effective in absorbing UVA (320-400 nm) and some UVB (280-320 nm) radiation.
  • Delays photodegradation of materials like silicones
  • Improves UV aging resistance
  • Enhances the photoprotective capabilities of sunscreen products
  • Intended for research use only
Catalog No. 50-004-08663