Learn More
Selleck Chemical LLC Bisoctrizole S4551-100mg
SDP
Supplier:  Selleck Chemical LLC S4551100MG
Encompass_Preferred
Bisoctrizole (Tinosorb M Tinuvin-360 UV-360 Milestab-360) is a broad-spectrum ultraviolet radiation absorber absorbing UVB as well as UVA rays Also a hybrid UV absorber reflecting and scattering UV
Catalog No. 50-311-1492