Learn More
Medchemexpress LLC Bisoprolol | 66722-44-9 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129029100MG
Encompass_Preferred
Bisoprolol is a potent, selective, and orally active β1-adrenergic receptor blocker with minimal activity on the β2-receptor. It is being researched for its potential use in hypertension, coronary artery disease, and stable ventricular dysfunction.
  • Potent, selective, and orally active β1-adrenergic receptor blocker
  • Minimal activity on β2-receptors
  • Researched for hypertension, coronary artery disease, and stable ventricular dysfunction
  • Protects myocardial cells from ischemia/reperfusion injury
  • Shows protective effects against cadmium-induced myocardial toxicity
  • Reverses small conductance calcium-activated potassium channel remodeling
Catalog No. 50-003-40439