Learn More
Apexbio Technology LLC Bisoprolol fumarate 104344-23-2 100mg
SDP
Supplier:  Apexbio Technology LLC B1354100
Encompass_Preferred
Bisoprolol fumarate (CAS 104344-23-2) is a selective 1-adrenergic receptor antagonist that preferentially inhibits 1-adrenergic signaling with minimal activity at 2 receptors By suppressing 1 receptor-mediated pathways it reduces both heart rate and myocardial contractility following oral administration The compound is utilized in pharmacological and physiological studies to investigate mechanisms underlying hypertension coronary artery disease and stable ventricular dysfunction as well as to evaluate therapeutic interventions that target sympathetic nervous system modulation in cardiovascular research contexts
Catalog No. 50-224-0801