Learn More
Selleck Chemical LLC Bisoprolol fumarate 25mg 104344-23-2 EMD33512
SDP
Supplier:  Selleck Chemical LLC S120625MG
Encompass_Preferred
Bisoprolol fumarate (EMD33512) is a selective type ?1 adrenergic receptor blocker. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1023