Learn More
Medchemexpress LLC Bisoprolol hemifumarate | 104344-23-2 | 99.8% | C18H31NO4.1/2C4H4O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0076500MG
Encompass_Preferred
Bisoprolol hemifumarate is a potent, selective, and orally active β1-adrenergic receptor blocker with little activity on β2-receptor. It has potential for research in hypertension, coronary artery disease, and stable ventricular dysfunction.
  • Protects myocardial cells from ischemia/reperfusion injury
  • Reduces H/R-induced ROS production and apoptosis
  • Increases AKT and GSK3β phosphorylation in H9c2 cells
  • Reverses Epinephrine-inhibited emigration in cholesterol-loaded DCs
  • Increases left ventricular ejection fraction and decreases heart rate
  • Shows protective effects against Cadmium-induced myocardial toxicity in rats
  • Reverses small conductance calcium-activated potassium channel remodeling in a volume-overload rat model
Catalog No. 50-004-32165