Learn More
Medchemexpress LLC Bisphenol A (3-chloro-2-hydroxypropyl)glycidyl ether | 13836-48-1 | MFCD10567366 | ≥90.0% | 376.87 g/mol | C21H25ClO4 | 1 MG

Supplier: Medchemexpress LLC HYW5858561MG
Bisphenol A (3-chloro-2-hydroxypropyl)glycidyl ether is a chlorinated bisphenol A derivative used as a synthetic intermediate in research and chemical synthesis. Manufacturer documentation provides identifying information and recommends storing the material under conditions stated in the certificate of analysis.
- Used as a drug intermediate and research chemical.
- CAS number 13836-48-1.
- Molecular formula C21H25ClO4; molecular weight 376.87 g/mol.
- Purity ≥90.0% (where specified by supplier analysis).
- For research use only; not for diagnostic or therapeutic use.
- May act as an endocrine disruptor; follow appropriate safety precautions.
- Store under conditions recommended in the certificate of analysis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.