Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bisphenol A (3-chloro-2-hydroxypropyl)glycidyl ether | 13836-48-1 | MFCD10567366 | >95.0% | 376.87 g/mol | C21H25ClO4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW5858565MG

Encompass_Preferred

Bisphenol A (3-chloro-2-hydroxypropyl)glycidyl ether is a chlorinated derivative of bisphenol A used as an analytical reference standard and intermediate in studies of food-contact materials, coatings, and chemical migration. It is applied in laboratory analysis and impurity profiling where identification of chlorinated glycidyl ethers is required.

  • Used as an analytical reference standard for migration and impurity studies.
  • Relevant chlorinated glycidyl ether structure for food-contact analysis.
  • Molecular formula C21H25ClO4 and molecular weight 376.87 g/mol.
  • CAS number 13836-48-1 for precise substance identification.
  • Supplied in small sample quantities suitable for analytical testing.

Catalog No. 50-002-84680


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.