Learn More
Medchemexpress LLC Bisphenol A bissulfate disodium | 10040-44-5 | MFCD22989096 | >95.0% | 432.38 | C15H14Na2O8S2 | 25 MG

Supplier: Medchemexpress LLC HYW58751625MG
Bisphenol A bissulfate disodium is the disodium salt metabolite of Bisphenol A supplied for research use. It is reported to exhibit complement-inhibiting activity (IC50 = 247 μM) and is typically used in in vitro biochemical assays and as a reference standard. The material is a white solid with molecular formula C15H14Na2O8S2 and molecular weight 432.38.
- Reported complement-inhibiting activity (IC50 247 μM).
- Molecular formula C15H14Na2O8S2; molecular weight 432.38.
- Provided as a white solid; typical package 25 MG.
- Purity >95% by HPLC as reported in standards documentation.
- Suitable for in vitro biochemical assays and reference analyses.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.