Learn More
Medchemexpress LLC Bisphenol A-d14 | 120155-79-5 | 99.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18260S610MG
Encompass_Preferred
Bisphenol A-d14 is a deuterium-labeled analytical standard of bisphenol A intended for research and quantitative analytical applications. It is used as an internal standard for mass spectrometry and other analytical methods that require isotopically labeled reference material.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High chemical purity (99.7%) suitable for analytical work.
  • Available in 10 mg size for small-scale analyses.
  • Chemical formula C15H2D14O2 and molecular weight 242.37 facilitate accurate calculations.
  • Supplied as a solid analytical standard for use as an internal reference.
Catalog No. 50-002-94871