Learn More
Medchemexpress LLC Bisphenol A-d14 | 120155-79-5 | 99.7% | 242.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY18260S65MG
Encompass_Preferred
Bisphenol A-d14 is a deuterium-labeled Bisphenol A. Bisphenol A is a phenolic, organic synthetic compound widely used in the production of polycarbonate plastics and epoxy resins. It is classified as an endocrine-disrupting compound (EDC) and is associated with various diseases, including cardiovascular diseases, respiratory diseases, diabetes, kidney diseases, obesity, and reproductive disorders. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules for quantitation during drug development. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87586