Learn More
Medchemexpress LLC Bisphenol A-d8 | 92739-58-7 | MFCD00143778 | 236.34 | C15H8D8O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18260S310MG
Encompass_Preferred
Bisphenol A-d8 is the deuterium-labeled isotopologue of bisphenol A used as an analytical reference and internal standard for quantitative mass spectrometry. It is intended for trace-level quantification and method development in environmental, food, and biological sample analysis.
  • Deuterium-labeled (d8) to enable isotope-based discrimination in mass spectrometry.
  • Provides a consistent internal standard for accurate quantification of bisphenol A.
  • Suitable for method development, validation, and trace-level analysis.
  • Chemical formula C15H8D8O2 and molecular weight 236.34 for reference use.
  • Packaged in small quantities appropriate for analytical standards (10 mg).
Catalog No. 50-002-95264