Learn More
Medchemexpress LLC Bisphenol AF-d4 | 2708286-36-4 | 100.0% | 340.25 | C15H6D4F6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW65407710MG
Encompass_Preferred
Bisphenol AF-d4 is the deuterium-labeled isotopologue of Bisphenol AF supplied for analytical and research use as a tracer and internal standard. Supplied as a 10 mg solid, it is intended for quantitative method development and accurate measurement of Bisphenol AF in biological and environmental samples using spectroscopic and mass spectrometric techniques.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High isotopic and chemical purity for reliable quantitation.
  • Compatible with NMR, GC-MS, and LC-MS workflows.
  • Supplied in a small vial appropriate for analytical-scale use.
Catalog No. 50-002-84381