Learn More
Medchemexpress LLC Bisphenol B-d8 | 2469735-90-6 | 98.9% | 250.36 | C16H10D8O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW6539751MG
Encompass_Preferred
Bisphenol B-d8 is a deuterium-labeled reference standard designed for use as a tracer or internal standard in quantitative analytical methods. Supplied as a high-purity isotopically labeled powder (98.9% purity) in a 1 mg quantity, it supports accurate measurement in NMR, GC-MS, and LC-MS workflows. Store powder at -20°C for long-term stability.
  • High isotopic labeling enables precise mass spectrometric quantitation.
  • High purity (98.9%) reduces background interference in analyses.
  • Compact 1 mg size suitable for trace-level standard preparation.
  • Stable when stored as powder at -20°C for extended shelf life.
Catalog No. 50-002-84848