Learn More
Medchemexpress LLC Bisphenol Z-d6 | 2733972-12-6 | 274.39 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW013326S1MG
Encompass_Preferred
Bisphenol Z-d6 is the deuterium labeled 4,4'-(Cyclohexane-1,1-diyl)diphenol. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-54663