Learn More
Strem, An Ascensus Company CAS 3385-94-2. 5g. Bis(trimethylsilyl)sulfide min. 98%. MFCD00014851
SDP
Supplier:  Strem, An Ascensus Company 1410155G
Encompass
CAS 3385-94-2. 5g. Bis(trimethylsilyl)sulfide min. 98%. MFCD00014851. Molecular Weight 178.44. Molecular Formula (CH3)3SiSSi(CH3)3. Color/form colorless liq. Strem 14-1015. http//www.strem.com/catalog/v/14-1015/
Catalog No. 50-211-0712