Learn More
Cayman Chemical BIsubstrate InhIbItr 78 5mg
SDP
Supplier:  Cayman Chemical 285675 MG
Encompass_Preferred
An NNMT inhibitor (IC50 = 1.41 µM); binds the NNMT active site in the binding pockets for SAM and nicotinamide; selective for NNMT over NSD2 and PRMT1 (IC50s = >50 µM for both); reduces MNA levels in, and inhibits proliferation of, HSC-2 oral cancer cells at 100 µM
Catalog No. 50-294-0746