Learn More
Selleck Chemical LLC Bitopertin
SDP
Supplier:  Selleck Chemical LLC S82195MG
Encompass_Preferred
Bitopertin (RG1678 RO-4917838) is a potent inhibitor of glycine transporter 1 (GlyT1) with Ki of 8 1 nM for human hGlyT1b and IC50 of 22-25 nM in Chinese hamster ovary cells
Catalog No. 50-217-3349