Learn More
Medchemexpress LLC Bivamelagon | 2641595-54-0 | C35H53ClN4O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15366010MM/1ML
Encompass_Preferred
Bivamelagon is an orally active MC4R agonist, characterized by an EC50 of 0.562 nM in the Luci assay and 36.5 nM in the cAMP assay, with a Ki of 65 nM. This compound is suitable for research into conditions such as obesity and diabetes.
  • Orally active MC4R agonist
  • Effective for research in obesity and diabetes
  • Exhibits β-arrestin recruitment activity
  • Demonstrates weight-loss activity in mouse obesity models
  • High solubility in DMSO
  • Purity of 98.72%
Catalog No. 50-003-73724