Learn More
Selleck Chemical LLC BIX 01294 10mg 1392399-03-9
SDP
Supplier:  Selleck Chemical LLC S800610MG
Encompass_Preferred
BIX01294 is an inhibitor of G9a histone methyltransferase with IC50 of 2.7 ?M in a cell-free assay, reduces H3K9me2 of bulk histones, also weakly inhibits GLP (primarily H3K9me3), no significant activity observed at other histone methyltransferases. BIX01294 induces autophagy. BIX01294 also inhibits H3K36 methylation by oncoproteins NSD1, NSD2 and NSD3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5541