Learn More
AdipoGen BIX 01294 . trihydrochloride,Chemical. CAS: 13923999-03-9
SDP
Supplier:  AdipoGen AGCR10150M025
Encompass_Preferred
BIX 01294 . trihydrochloride, AdipoGen Life Sciences. Chemical. CAS: 1392399-03-9 | 935693-62-2 (free base). Formula: C28H38N6O2 . 3HCl. MW: 490.6 . 109.4. Selective and cell permeable G9a histone-lysine methyltransferase (HMTase) inhibitor. Does not compete with cofactor S-adenosylmethionine. Pluripotent stem cell inducer.
Catalog No. 50-114-6508